Learn More
Macherey-Nagel™ Quantofix™ Peracetic Acid 50 Test Strip Kits
Allows for the quick and easy determination of peracetic acid in solutions. Macherey Nagel™ Quantofix™ Peracetic Acid 50 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand: Macherey-Nagel™ 91340
Description
Allows for the quick and easy determination of peracetic acid in solutions. Macherey Nagel™ Quantofix™ Peracetic Acid 50 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Ready to use
- Rapid: provides reliable results within 30 seconds
- Excellent for checking disinfected bottles in the beverage industry and dialysis machines
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store under 30°C
Specifications
Peracetic Acid 50 Test Strip | |
0 to 50mg/L | |
Semi Quantitative Test Strips |
For the fast and reliable determination of peracetic acid in solutions | |
100 Pack |
Product Suggestions
Customers who viewed this item also viewed.