missing translation for 'onlineSavingsMsg'
Learn More
Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene), Thermo Scientific Chemicals Product Code.: 11323786

Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene), Thermo Scientific Chemicals

Product Code. 11323786
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11323786

Brand: Thermo Scientific Alfa Aesar L16836.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2070-70-4
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00153253
InChI Key SAPOZTRFWJZUFT-UHFFFAOYSA-N
Synonym 2z-1,1,1,2,3,4,5,5,5-nonafluoro-4-trifluoromethyl pent-2-ene
PubChem CID 11012007
SMILES FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Linear Formula (CF3)2CFCF=CFCF3
Refractive Index <1.3000
Quantity 100 g
IUPAC Name (Z)-1,1,1,2,3,4,5,5,5-nonafluoro-4-(trifluoromethyl)pent-2-ene
Formula Weight 300.05
Percent Purity ≥90%
Assay contains 5 to 10% perfluoro(2-methyl-2-pentene)
Chemical Name or Material Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1]

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10203483 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberL16836.14,L16836.22
Lot Number10204314 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberL16836.14,L16836.22
Lot Number10204314 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16836.14,L16836.22
Lot Number10174669 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16836.14,L16836.22
Lot Number10203483 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL16836.14,L16836.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.