missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PeridoxRTU
BIOPHARMA

PeridoxRTU is a blend of hydrogen peroxide and peracetic acid.

Brand:  Contec™ FBC3702PX

177.63 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.



 View more versions of this product

Product Code. 17251502

  • £212.00 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contec PeridoxRTU is a BPR and EPA authorised sporicide achieving greater than a log 3 reduction against bacterial and fungal spores in under 3 min
  • A blend of hydrogen peroxide and peracetic acid, Contec PeridoxRTU is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts and spores
  • PeridoxRTU's proprietary formulation contains surfactants for cleaning efficiency along with corrosion inhibitors for surface protection
  • PeridoxRTU can be used to clean and disinfect surfaces without damage or loss of functionality
  • Contec PeridoxRTU is filtered and filled in a controlled environment
  • The sterile product is provided validated sterile to a Sterility Assurance Level of 10-6
  • Provided double or single bagged, the product is designed for ease of entry into pharmaceutical cleanrooms
  • Available as either a 0.9 L bottle with a push/pull cap for small areas or in 3.78 L capped containers for mop head saturation or when larger volumes are required
  • Patented hyperactive chemistry makes it fast acting so save time spent of biodecontamination
  • Validated sterile version available makes it suitable for Grade A and B cleanrooms
  • Excellent material compatibility and will not damage common cleanroom materials
  • Double/single bagged helps facilitate transfer disinfection into cleanroom
  • Push/pull cap on 0.9 L eliminates droplets reducing the risk of inhalation and can be easily applied to a wipe or cleaning too
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner
Not Applicable
Non-sterile
Not Autoclavable
Hydrogen Peroxide and Peracetic Acid
Limited-use
No
Liquid
Unscented
Clear
Clear
RTU
Non-filtered
3.78 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X4 Peridox Ready to Use Sporicidal Disinfectant, 3

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberGH32080801231C Certificate TypeCertificate of Quality Date26/01/2024 Catalog NumberFBC3702PX
Lot NumberGH32080412231D Certificate TypeCertificate of Quality Date20/10/2023 Catalog NumberFBC3702PX
Lot NumberGH32081213221F Certificate TypeCertificate of Quality Date01/09/2023 Catalog NumberFBC3702PX
Lot NumberGH32080826221A Certificate TypeCertificate of Quality Date06/01/2023 Catalog NumberFBC3702PX
Lot NumberGH32080802201D Certificate TypeCertificate of Compliance Date11/03/2022 Catalog NumberFBC3702PX
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Contec™ PeridoxRTU > Volume: 3.78 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.