Learn More
Thermo Scientific™ Blocker™ BSA
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Brand: Thermo Scientific™ 37525
211.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.
Specifications
Blocker BSA in PBS (10X) | |
200 mL | |
Western Blot, ELISA, Immunohistochemistry (IHC) | |
10% BSA | |
Single purified protein |
Blocker BSA | |
Liquid | |
Blocker™ | |
PBS | |
Store at 4°C. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZL403846 | Certificate of Analysis | 30/01/2025 | 37525_LITL18 PROMO,37525 |
ZK403514 | Certificate of Analysis | 16/12/2024 | 37525_LITL18 PROMO,37525 |
ZK403515 | Certificate of Analysis | 16/12/2024 | 37525_LITL18 PROMO,37525 |
ZJ402787 | Certificate of Analysis | 16/12/2024 | 37525_LITL18 PROMO,37525 |
ZJ402788 | Certificate of Analysis | 16/12/2024 | 37525_LITL18 PROMO,37525 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.