Learn More
Brady™ PermaSleeve™ Yellow Heat Shrink Wire and Cable Labels for M6/M7 Printers
Designed for rigorous wire marking applications, the PermaSleeve™ Heat Shrink Polyolefin labels combine speed, ease of use, and permanence to create high quality labeling.
£54.56 - £200.17
Specifications
Certifications/Compliance | ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) |
---|---|
Adhesive Type | Non-adhesive |
Color | Yellow |
Finish | Matte |
Format | Die-cut |
Product Code | Brand | Diameter (English) | Dimensions (LxW) | Wire Gauge | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Dimensions (LxW) | Wire Gauge | For Use With (Equipment) | Price | Quantity & Availability | ||
17849214
|
Brady™
174653 |
0.094 in. | 0.182 x 1 in. | 28 to 20 AWG | M710, BMP71 |
£60.45
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17841409
|
Brady™
173589 |
0.094 in. | 0.182 x 1 in. | 28 to 20 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£54.56
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17888934
|
Brady™
174655 |
0.094 in. | 0.182 x 1.75 in. | 28 to 20 AWG | M710, BMP71 |
£78.55
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17851429
|
Brady™
173583 |
0.094 in. | 0.182 x 1.75 in. | 28 to 20 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£74.47
Pack of 100 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17869184
|
Brady™
174541 |
0.125 in. | 0.235 x 1 in. | 22 to 16 AWG | M710, BMP71 |
£62.50
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17821469
|
Brady™
173614 |
0.125 in. | 0.235 x 1 in. | 22 to 16 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£59.23
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17821539
|
Brady™
173609 |
0.125 in. | 0.235 x 1.75 in. | 22 to 16 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£65.11
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17829204
|
Brady™
174545 |
0.125 in. | 0.235 x 1.75 in. | 22 to 16 AWG | M710, BMP71 |
£81.15
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17849074
|
Brady™
174560 |
0.187 in. | 0.335 x 1 in. | 20 to 10 AWG | M710, BMP71 |
£63.65
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17811639
|
Brady™
173630 |
0.187 in. | 0.335 x 1 in. | 20 to 10 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£60.36
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17808764
|
Brady™
174564 |
0.187 in. | 0.335 x 1.75 in. | 20 to 10 AWG | M710, BMP71 |
£82.75
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17861429
|
Brady™
173623 |
0.187 in. | 0.335 x 1.75 in. | 20 to 10 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£74.75
Pack of 100 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17818964
|
Brady™
174579 |
0.25 in. | 0.439 x 1 in. | 16 to 8 AWG | M710, BMP71 |
£72.15
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17898754
|
Brady™
174583 |
0.25 in. | 0.439 x 1.75 in. | 16 to 8 AWG | M710, BMP71 |
£93.75
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17861399
|
Brady™
173644 |
0.25 in. | 0.444 x 1 in. | 16 to 8 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£68.40
Pack of 100 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17861579
|
Brady™
173638 |
0.25 in. | 0.444 x 1.75 in. | 16 to 8 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£88.90
Pack of 100 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17831699
|
Brady™
173660 |
0.375 in. | 0.645 x 1 in. | 12 to 4 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£70.61
Pack of 100 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17809004
|
Brady™
174611 |
0.375 in. | 0.645 x 1 in. | 12 to 4 AWG | M710, BMP71 |
£78.20
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17851389
|
Brady™
173654 |
0.375 in. | 0.645 x 1.75 in. | 12 to 4 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£96.35
Pack of 100 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17868764
|
Brady™
174615 |
0.375 in. | 0.645 x 1.75 in. | 12 to 4 AWG | M710, BMP71 |
£102.00
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17861489
|
Brady™
173672 |
0.5 in. | 0.851 x 1 in. | 8 to 1 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£85.73
Pack of 100 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17839114
|
Brady™
174628 |
0.5 in. | 0.851 x 1 in. | 8 to 1 AWG | M710, BMP71 |
£90.40
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17881569
|
Brady™
173669 |
0.5 in. | 0.851 x 1.75 in. | 8 to 1 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£106.11
Pack of 100 |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17899074
|
Brady™
174632 |
0.5 in. | 0.851 x 1.75 in. | 8 to 1 AWG | M710, BMP71 |
£118.00
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17801479
|
Brady™
173679 |
0.75 in. | 1.25 x 1 in. | 6 to 0000 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£153.95
Pack of 100 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17811399
|
Brady™
173677 |
0.75 in. | 1.25 x 1.75 in. | 6 to 0000 AWG | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£200.17
Pack of 100 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17881509
|
Brady™
173602 |
1 in. | 1.667 x 1 in. | 2 to 500 MCM | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£70.31
Pack of 50 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17889024
|
Brady™
174524 |
1 in. | 1.667 x 1 in. | 2 to 500 MCM | M710, BMP71 |
£77.85
Pack of 50 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17851369
|
Brady™
173600 |
1 in. | 1.667 x 1.75 in. | 2 to 500 MCM | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) |
£91.41
Pack of 50 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17828834
|
Brady™
174528 |
1 in. | 1.667 x 1.75 in. | 2 to 500 MCM | M710, BMP71 |
£102.00
Pack of 50 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
Description
- Industrial grade with high temperature, UV, chemical, fuel, and oil resistance
- Self-extinguishing labels
- Crisp, clear identification resistant to abrasions and fading
- 3:1 heat shrinkable labels fit snugly around wires for maximum insulation protection and permanent identification
- Made of B-342 polyolefin material
Specifications
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) | |
Yellow | |
Die-cut | |
Labels | |
Electrical Labeling, Wire and Cable Labeling | |
-67°F to +275°F |
Non-adhesive | |
Matte | |
Polyolefin | |
Non-sterile | |
Rectangle | |
-55°C to +135°C |