missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Permeabilization Medium (Medium B)

Permeabilization Medium (Medium B)

£72.25 - £584.00

Specifications

Content And Storage Room Temperature
Type Permeabilization Medium (Medium B)
Reagent Type Permeabilization Reagent
Shipping Condition Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10314023
View Documents
Invitrogen™
GAS002S5
5 mL
£72.25
Each
Please to purchase this item. Need a web account? Register with us today!
10572815
View Documents
Invitrogen™
GAS002S100
100 mL
£584.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Permeabilization Medium (Medium B) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.

The FIX & PERM™ Cell Permeabilization Kit is:

  • Flexible—compatible with multiple sample types and cellular antigens
  • Robust—scatter pattern of cells unaffected
  • Efficient—simultaneous detection of intracellular and surface markers

    View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).

    The Permeabilization Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.

    How It Works
    Cells are incubated for 15 minutes at room temperature with Fixation Medium (Medium A) (Cat. No. GAS001S100) and washed. Cells are then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
  • Specifications

    Specifications

    Room Temperature
    Permeabilization Medium (Medium B)
    Permeabilization Reagent
    Room Temperature
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Invitrogen™ Permeabilization Medium (Medium B)

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.