Learn More
Christeyns™ Phago'Derm Sensitive SPS Soap
Phago'Derm Sensitive SPS Soap is intended for simple washing of hands and forearms in intensive use. It preserves the protective barrier of the skin and provides a feeling of comfort.
Brand: Christeyns™ C65085
Additional Details : Weight : 10.69010kg
Description
- Does not dry out the skin, even with frequent use
- Hypoallergenic (formulated to minimize allergic reactions)
- Suitable for washing children under 36 months and pregnant/breastfeeding women
- Suitable for patient washing and preoperative showering
- Good cleaning power.
- High rinsability
- Hydrating potential for 30 min.
Specifications
Hand Soap | |
Foam | |
1000 mL |
Colorless | |
12 Airless Mounted Pump Bottles |
Safety and Handling
- 12LT X Soap PHAGODERM SENSITIVE SPS 1LT bottle Pum
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.