missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Phenolphthalein, 98% Product Code.: 11498996

Thermo Scientific Chemicals Phenolphthalein, 98%

Artikelnummer. 11498996
50 g
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1000 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Product Code. 11498996

Brand: Thermo Scientific Chemicals A17135.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

Solubility
Soluble in alcohol. Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.

Notes
Acid-base indicator: pH 8.0 - 10.0.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Spezifikation

Melting Point 258°C to 262°C
Density 1.299
Quantity 50 g
Beilstein 284423
Merck Index 14,7243
Solubility Information Soluble in alcohol. Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Formula Weight 318.33
Percent Purity 98%
Chemical Name or Material Phenolphthalein

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249131 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog NumberA17135.0B,A17135.30,A17135.18
Lot Number10239045 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17135.0B,A17135.30,A17135.18
Lot Number10190954 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17135.0B,A17135.30,A17135.18
Lot NumberD4342 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17135.0B,A17135.30,A17135.18
Lot Number61200017 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17135.0B,A17135.30,A17135.18
compliance-icons
Produktidentifikator
  • Phenolphthalein
Signalwort
  • Danger
Gefahrenkategorie
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Gefahrenhinweis
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Phenolphthalein

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Thermo Scientific Chemicals Phenolphthalein, 98% >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt