missing translation for 'onlineSavingsMsg'
Learn More
Phenothiazine, 99%, Thermo Scientific Chemicals Product Code.: 10214800

Phenothiazine, 99%, Thermo Scientific Chemicals

Product Code. 10214800
500 g, Plastic Bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10214800

Brand: Thermo Scientific Acros 130215000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-84-2
Molecular Formula C12H9NS
Molecular Weight (g/mol) 199.28
MDL Number MFCD00005015
InChI Key WJFKNYWRSNBZNX-UHFFFAOYSA-N
Synonym phenothiazine, thiodiphenylamine, feeno, dibenzothiazine, phenosan, phenthiazine, dibenzo-1,4-thiazine, penthazine, souframine, agrazineShow MoreShow Less
PubChem CID 7108
ChEBI CHEBI:37931
IUPAC Name 10H-phenothiazine
SMILES C1=CC=C2C(=C1)NC3=CC=CC=C3S2

Specifications

Melting Point 185.0°C
Color Yellow
Boiling Point 371.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic Bottle
Quantity 500 g
Beilstein 27, 63
Merck Index 15, 7364
Solubility Information Solubility in water: 2mg/l (25°C). Other solubilities: freely soluble in hot acetic acid, freely soluble in benzene and ether, slightly soluble in alcohol and mineral oils, practically insoluble in chloroform and petroleum, ether
Formula Weight 199.28
Percent Purity 99%
Physical Form Prills or Beads
Chemical Name or Material Phenothiazine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466796 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number130210050,130215000,130210010
Lot NumberA0463349 Certificate TypeCertificate of Analysis Date30/05/2024 Catalog Number130210050,130215000,130210010
Lot NumberA0458545 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number130210050,130215000,130210010
Lot NumberA0458546 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number130210050,130215000,130210010
Lot NumberA0455490 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number130210050,130215000,130210010
compliance-icons compliance-icons
Product Identifier
  • Phenothiazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenothiazine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.