missing translation for 'onlineSavingsMsg'
Learn More
Phloroglucinol, anhydrous, 98%, Thermo Scientific Chemicals Product Code.: 11427186

Phloroglucinol, anhydrous, 98%, Thermo Scientific Chemicals

Product Code. 11427186
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11427186

Brand: Thermo Scientific Alfa Aesar B25502.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .

Solubility
Soluble in diethyl ether, ethanol and pyridine.

Notes
Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-73-6
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00002286
InChI Key QCDYQQDYXPDABM-UHFFFAOYSA-N
Synonym phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxyShow MoreShow Less
PubChem CID 359
ChEBI CHEBI:16204
IUPAC Name benzene-1,3,5-triol
SMILES C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 215°C to 220°C
Quantity 250 g
Beilstein 1341907
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,7328
Solubility Information Soluble in diethyl ether,ethanol and pyridine.
Formula Weight 126.11
Percent Purity 98%
Chemical Name or Material Phloroglucinol, anhydrous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ07J025 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog NumberB25502.18,B25502.30
Lot NumberM31K031 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog NumberB25502.18,B25502.30
Lot Number10244426 Certificate TypeCertificate of Analysis Date03/11/2022 Catalog NumberB25502.18,B25502.30
Lot Number5011A14W Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25502.18,B25502.30
Lot Number10098279 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25502.18,B25502.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phloroglucinol, anhydrous, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.