Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
Used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells.
Brand: Thermo Scientific Chemicals J61917.K2
68.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula is used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells. It plays an important role in the transportation of tissue samples and as diluents in various reagent preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Ambient temperatures | |
Plastic bottle | |
1 L | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M16L515 | Certificate of Analysis | 24/01/2025 | J61917.K2,J61917.K3,J61917.AP |
Z28K536 | Certificate of Analysis | 21/01/2025 | J61917.K2,J61917.K3,J61917.AP |
X28K518 | Certificate of Analysis | 19/11/2024 | J61917.K2,J61917.K3,J61917.AP |
U25K507 | Certificate of Analysis | 18/09/2024 | J61917.K2,J61917.K3,J61917.AP |
U06K502 | Certificate of Analysis | 16/08/2024 | J61917.K2,J61917.K3,J61917.AP |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only