missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphate-Citrate Buffer (5X)

Used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.

Brand:  Thermo Scientific™ J62982.AE

58.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15415269

  • £59.25 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
Plastic Bottle
Ambient Temperatures
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP06L007 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberJ62982.AE
Lot NumberN21K014 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberJ62982.AK,J62982.AE
Lot NumberX20J015 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberJ62982.AK,J62982.AE
Lot NumberN28I038 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog Number
Lot NumberX28I021 Certificate TypeCertificate of Analysis Date27/11/2022 Catalog Number
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Phosphate-Citrate Buffer (5X) > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.