Learn More
Phosphorus pentasulfide, 98+%, Thermo Scientific Chemicals
£92.16 - £142.71
Chemical Identifiers
CAS | 1314-80-3 |
---|---|
Molecular Formula | P4S10 |
Molecular Weight (g/mol) | 444.48 |
MDL Number | MFCD00011441 |
InChI Key | CYQAYERJWZKYML-UHFFFAOYSA-N |
Synonym | phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech |
PubChem CID | 14817 |
SMILES | P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10057682
|
Thermo Scientific Acros
196725000 |
500 g | Glass bottle |
£92.16
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703011
|
Thermo Scientific Acros
196720010 |
1 kg | Glass bottle |
£142.71
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1314-80-3 | |
444.48 | |
CYQAYERJWZKYML-UHFFFAOYSA-N | |
14817 |
P4S10 | |
MFCD00011441 | |
phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech | |
P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3 |
Specifications
514.0°C | |
1314-80-3 | |
Powder | |
P4S10 | |
MFCD00011441 | |
>100°C | |
phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech | |
CYQAYERJWZKYML-UHFFFAOYSA-N | |
444.48 | |
444.48 | |
2.08 |
282.0°C to 286.0°C | |
Yellow to Green | |
98% min | |
P4S10 | |
01,870; 03,226; 04,389; 05,534; 06,470; 07,290; 08,401; 09,374; 10,320 | |
15,7466 | |
Solubility in water: reacts forming h3po4 and h2s. Other solubilities: soluble in solutions of alkali hydroxides, soluble in carbon disulfide, reacts with alcohols and acids | |
P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3 | |
14817 | |
≥98% | |
Phosphorus pentasulfide |
Safety and Handling
GHS H Statement
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.
Contact with water liberates toxic
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-242-4
- Phosphorus pentasulfide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H228-Flammable solid.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.