missing translation for 'onlineSavingsMsg'
Learn More
Phthalide-3-acetic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11322316

Phthalide-3-acetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11322316
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11322316

Brand: Thermo Scientific Alfa Aesar L05984.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4743-58-2
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00051729
InChI Key FJWKEFBYCZSVNZ-UHFFFAOYSA-N
Synonym phthalide-3-acetic acid, 3-oxo-1,3-dihydro-2-benzofuran-1-yl acetic acid, 2-3-oxo-1,3-dihydroisobenzofuran-1-yl acetic acid, 3-oxo-1,3-dihydro-isobenzofuran-1-yl-acetic acid, 2-3-oxo-1,3-dihydro-2-benzofuran-1-yl acetic acid, 3-oxo-1h-2-benzofuran-1-yl acetic acid, 2-3-oxo-1h-2-benzofuran-1-yl acetic acid, 2-3-oxohydroisobenzofuranyl acetic acid, acmc-20ajvy, 1,3-dihydro-3-oxo-1-isobenzofuranacetic acidShow MoreShow Less
PubChem CID 589730
IUPAC Name 2-(3-oxo-1H-2-benzofuran-1-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(OC2=O)CC(=O)O

Specifications

Melting Point 148°C to 153°C
Quantity 25 g
Beilstein 156005
Formula Weight 192.17
Percent Purity ≥98%
Chemical Name or Material Phthalide-3-acetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10112645 Certificate TypeCertificate of Analysis Date27/07/2023 Catalog NumberL05984.06,L05984.14
Lot Number10112645 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05984.06,L05984.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phthalide-3-acetic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.