missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Human Specimen Direct PCR Kit

Thermo Scientific Phusion Human Specimen Direct PCR Kit delivers unprecedented convenience for DNA amplification by allowing PCR directly from unpurified human samples.

Brand:  Thermo Scientific™ F150BID

Product Code. 10639209

  • £312.00 / 200 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Phusion Human Specimen Direct PCR Kit delivers unprecedented convenience for DNA amplification by allowing PCR directly from unpurified human samples. A tiny amount of sample is used directly in the PCR reaction with no prior purification steps. This allows significant savings in both time and cost.

Tested sample materials include buccal swabs, saliva, amniotic fluid, hair, fingernails, teeth, and skin biopsies. The kit is compatible with fresh, frozen, and even formalin-fixed, paraffin-embedded (FFPE) human tissue samples. The Phusion Human Specimen Direct PCR Kit includes optimized reagents and detailed protocols for many sample materials. The modified Phusion Hot Start II High-Fidelity DNA Polymerase utilized in the kit is a highly accurate proofreading DNA polymerase, and PCR products obtained are perfectly suitable even for cloning purposes.

Features

  • No need for time-consuming and expensive DNA purification steps
  • Two simple protocols for various applications
  • Extremely short PCR protocol times
  • Modified Phusion Hot Start II High-Fidelity DNA Polymerase delivers abundant yields of specific product

Notes

  • The optimal annealing temperature for Phusion DNA Polymerases may differ significantly from that of Taq-based DNA polymerases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Phusion Human Specimen DNA Polymerase, 80 μL
• 2X Phusion Human Specimen PCR Buffer, 2 x 1.25 mL
• Universal Control Primer Mix (25 μM), 40 μL
• Dilution Buffer, 2 x 5 mL
• DNARelease Additive, 3 x 100 μL

Store at -20°C.
Direct PCR
Fast
200 reactions
Hot-start PCR
200 reactions
Separate Components
DNA
High
Phusion Human Specimen DNA Polymerase
Direct PCR Kit
Wet or Dry Ice
Built-In Hot Start
Blunt
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Reference Materials

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3081819 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberF150BID
Lot Number3017052 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberF150BID
Lot Number3130967 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberF150BID
Lot Number3081913 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberF150BID
Lot Number2872638 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberF150BID
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Phusion Human Specimen Direct PCR Kit > 200 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.