missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Phytone™ Peptone

An animal origin-free (AOF) enzymatic digest of soy.

Brand:  Gibco™ 211906

 View more versions of this product

Product Code. 16269761

  • £94.25 / 454g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • An animal origin-free (AOF) enzymatic digest of soy
  • A nutritious and excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
  • Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics
  • Also used with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines

Applications of Phytone Peptone include:

  • It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
  • It has been used in mammalian cell culture applications due to its high carbohydrate content
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LUO
Powder
Bottle
Phytone™
Animal origin-free, soy
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Dehydrated Culture Media
Production of monoclonal antibodies for oncological and rheumatological therapeutics, microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines
454 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3110387 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number211906
Lot Number2803431 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number211906
Lot Number2484059 Certificate TypeCertificate of Analysis Date16/05/2022 Catalog Number211906
Lot Number2448264 Certificate TypeCertificate of Analysis Date10/03/2022 Catalog Number211906
Lot Number2399853 Certificate TypeCertificate of Analysis Date02/11/2021 Catalog Number211906
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Phytone™ Peptone > 454 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.