missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Safety Shield
Safety Shield is free standing, portable and stables, with non-skid feet to provide facial protection from hazardous materials.
Brand: KARTELL 0057200
212.25 GBP valid until 2025-06-08
Use promo code "24131" to get your promotional price.
This item is not returnable.
View return policy
Description
- The safety shields are available in two sizes with different thicknesses and are made of high quality, clear acrylic; are free standing, portable and stable, and are all fitted with non -skid feet.
- Art. 571 is designed to provide laboratory and healthcare personnel with facial protection from hazardous substances, aerosol and splashing.
Specifications
18.9 in. | |
Rectangle | |
9 mm | |
305 mm | |
Acrylic | |
None |
480 mm | |
0.35 in. | |
12.01 in. | |
Countertop Shield | |
Reusable | |
No Frame |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KARTELL Safety Shield > Thickness: 9 mm
Spot an opportunity for improvement?Share a Content Correction