missing translation for 'onlineSavingsMsg'
Learn More
Piceatannol, Tocris Bioscience™ Product Code.: 30001375

Piceatannol, Tocris Bioscience™

Product Code. 30001375
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 30001375

Brand: Tocris Bioscience 1554/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits TNF-induced NF-κB activation

Anti-inflammatory, immunomodulatory and antiproliferative agent. Inhibits p56lck and syk protein tyrosine kinases and inhibits TNF-induced NF-κB activation and gene expression. Synthesis results from conversion of resveratrol (Cat. No. 1418) by cytochrome P450 1B1.
TRUSTED_SUSTAINABILITY

Specifications

CAS 10083-24-6
Molecular Formula C14H12O4
Quantity 10 mg
Synonym piceatannol, 3-hydroxyresveratol, astringinin, piceatanol, e-4-3,5-dihydroxystyryl benzene-1,2-diol, 3,3',4'5-tetrahydroxystilbene, 3,5,3',4'-tetrahydroxystilbene, 4-e-2-3,5-dihydroxyphenyl ethenyl benzene-1,2-diol, 3,3',4,5'-tetrahydroxystilbene, unii-6ks3ls0d4f
InChI Key CDRPUGZCRXZLFL-OWOJBTEDSA-N
SMILES C1=CC(=C(C=C1C=CC2=CC(=CC(=C2)O)O)O)O
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,2-diol
Molecular Weight (g/mol) 244.246
PubChem CID 667639
ChEBI CHEBI:28814
Formula Weight 244.25
Percent Purity >98%
Chemical Name or Material Piceatannol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Piceatannol, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.