missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 96-Well Polystyrene Plates, Corner Notch

Bind antibodies with high capacity using these specially formulated polystyrene plates for ELISA and other immunoassays.

Brand:  Thermo Scientific™ 15041

437.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10563364

  • £472.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics.

Pierce 96-Well Microplates are standard flat-bottom clear and white assay plates for ELISA and other protein-based methods. Each lot is certified to ensure adherence to strict performance criteria. Clear and opaque white varieties are available in 100-plate and 25-plate cases, respectively. Use the clear plates to perform colorimetric ELISA and other assays for measurement in standard spectrophotometer plate readers. Use the white plates to perform fluorescent or chemilumiescent assays for detection in fluorescence or luminometer readers.

Highlights:

  • Specially formulated optically clear polystyrene and treated to provide protein coating capacity
  • High binding capacity surface improves the overall performance in immunoassay applications
  • Certified for performance, having consistently low coefficients of variation (CV) among wells
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adsorption, Hydrophilic
Pierce 96-Well Polystyrene Plates, Corner Notch
100 Plate
Clear
Polystyrene
96
Flat Bottom (F-well)
100 plates
Store at room temperature.
Colorimetric
Pierce
Well Plate
360 μL
Polystyrene
None
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL4428931 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number15041
Lot NumberZL4428931A Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number15041
Lot NumberZK4428921 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number15041
Lot NumberSPECS Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number15041
Lot NumberZJ4323541 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number15041
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ 96-Well Polystyrene Plates, Corner Notch > Clear, corner notch

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.