Learn More
Thermo Scientific™ Pierce™ ChIP-grade Protein A/G Magnetic Beads
Shop All Thermo Scientific ProductsBeschreibung
Efficiently isolate chromatin-bound DNA by immunoprecipitation (chromatin IP) for subsequent quantitation by PCR using Thermo Scientific™ Pierce Magnetic ChIP Kit.
Additional Information and Online Ordering
Highlights:
Rapid – obtain purified DNA that is ready for quantitative PCR in about 8 hours
Efficient and reproducible – micrococcal nuclease digestion and nuclear lysis are highly optimized
Sensitive – obtain results with as few as 10,000 cells (1 x 104)
Low background – Pierce Protein A/G Magnetic Beads are blocked in a non-DNA-containing reagent to minimize background
Complete – optimized positive control reagents are included: RNA polymerase II antibody and GAPDH promoter PCR primers
Includes:
Kit contains reagents for chromatin preparation, IP, DNA purification, and positive controls (antibody and primers)
Requires:
In vivo crosslinker (such as formaldehyde), micro-tip sonicator (such as Misonix™ Sonicator 3000), ChIP qualified antibody of choice, PCR primers for DNA sequence of interest, PCR master mix (containing a dye such as SYBR™ Green, if qPCR is desired), and a qPCR instrument
Recommended for:
- Determine sites of specific protein-DNA interactions on genomic DNA by PCR or ChIP-Seq
- Monitor the effects of histone modifications or chemical agents on DNA binding
Spezifikation
Spezifikation
| Description | Pierce ChIP-grade Protein A/G Magnetic Beads |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area. |
| Product Line | Pierce |
| Quantity | 5 mL |
| Target | Antibody |
| Type | Magnetic Bead (ChIP grade) |
| Ligand Type | Protein A/G |
| Sufficient For | Approx. 250 Assays |
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.