missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Dilution-Free™ Rapid Gold BCA Protein Assay

The Pierce Dilution-Free Rapid Gold BCA Protein Assay Kit is an effortless, fast, and accurate assay optimized to determine total protein concentration. It is detergent-compatible and has a 5X wider dynamic range than traditional protein assays of 0.02–10 mg/mL.

Brand:  Thermo Scientific™ A55861

 View more versions of this product

Product Code. 17988192

  • £179.00 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

20 to 10,000 μg/mL
Colorimetric
Protein Quantitation Assay
Solution-based Detection, Absorbance
250 mL
Sufficient for: 1,200 microplate assays or 125 test-tube assays
• Dilution-Free Rapid Gold BCA Reagent A, 250 mL
• Dilution-Free Rapid Gold BCA Reagent B, 10 mL
• Dilution-Free BSA Protein Standards (0.2-10 mg/mL), 2 x 1 mL 8-channel tubestrip
Store in refrigerator (2°C to 8°C).
Pierce™ Dilution-Free™
Not Target-specific
Spectrophotometer, Microplate Reader
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK397019 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA55861
Lot NumberZK397019A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA55861
Lot NumberZG393910 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberA55861
Lot NumberZG393910A Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberA55861
Lot NumberZD383652 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog NumberA55861
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Dilution-Free™ Rapid Gold BCA Protein Assay > 250 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.