missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Glutathione Superflow Agarose

Perform high-purity, large-scale purification of GST-tagged proteins with glutathione that has been immobilized to a crosslinked 6% beaded agarose.

Brand:  Thermo Scientific™ 25236

193.12 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 13424199

  • £272.00 / 10mL
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.

Additional Information and Online Ordering

Pierce Glutathione Superflow Agarose is a highly crosslinked, durable resin that does not compress at flow rates required for medium- to large-scale FPLC purifications. The resin holds up well to a variety of chemical and pH values, and it is compatible with common clean-in-place procedures. The specific binding of glutathione to the GST-tag results in high purity and high yield in a single step purification process.

Highlights:

  • High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate

  • High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates

  • Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour

  • Compatible – maintains function after exposure to a wide variety of chemicals and pH values

  • Cost effective – resin is stable through multiple cycles of cleaning and reuse

Recommended for:

Medium- to large-scale FPLC purification of GST-tagged proteins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mg Pure GST Binding per mL Resin at 150 cm/hour Flow Rate, 20 mg per mL at 30 cm/hour Flow Rate
Pierce™
GST-Tagged Protein
Glutathione
Affinity, Agarose Resin
Upon receipt store at 4°C.
10 mL
Agarose
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.