missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MBP Column Preparation Buffer for Pierce™ IgM Purification Kit

Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.

Brand:  Thermo Scientific™ 21018

Product Code. 11896794

  • £59.50 / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce IgM Purification Kit is an easy-to-use affinity matrix and buffer system to purify IgM from mouse ascites fluid; the method uses immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.

Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5mL column of the MBP Agarose.

Highlights:

  • Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum

  • Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5mg IgM from 0.5mL ascites samples per column-use

  • Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies

  • Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity

Includes:

Kit contains a 5mL MBP Agarose Column, column preparation buffer, IgM binding and elution buffers, and a column extender

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgM Purification Kit
Conditioning Reagent
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C.
Liquid
Pierce™
Pierce MBP Column Preparation Buffer
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZH399395 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number21018
Lot NumberYJ379904 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number21018
Lot NumberXL358547 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number21018
Lot NumberXD346394 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog Number21018
Lot NumberWG329358 Certificate TypeCertificate of Analysis Date06/08/2021 Catalog Number21018
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MBP Column Preparation Buffer for Pierce™ IgM Purification Kit > MBP Column Preparation Buffer; 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.