Learn More
Thermo Scientific™ Pierce™ Iminobiotin Agarose
Agarose beads of immobilized biotin (and iminobiotin) to strongly (or reversibly) bind and remove (or purify) streptavidin or avidin proteins and conjugates from samples.
Brand: Thermo Scientific™ 20221
Description
Thermo Scientific™ Pierce Iminobiotin and Biotin Resins provide for unusual affinity purification experiments involving clean-up or removal of streptavidin or avidin protein components.
Pierce Biotin Agarose and Iminobiotin Agarose are beaded agarose supports onto which biotin and iminobiotin moieties have been covalently immobilized. Typical avidin-biotin affinity procedures use immobilized avidin or streptavidin proteins to capture biotin-tagged molecules from a sample. Occasionally, however, the reverse situation arises in which immobilized biotin in needed to capture avidin or streptavidin components from a sample. These biotin and iminobiotin resins address this special situation.Immobilized biotin binds avidin and streptavidin very strongly, making it appropriate for non-reversible binding applications. Immobilized iminobiotin is the guanido analog of biotin. The dissociation constant of the avidin-iminobiotin complex is pH dependent. At pH 9.5 to 11, the avidin-iminobiotin complex will bind tightly. At pH 4, the avidin-iminobiotin complex will dissociate. Thus, iminobiotin agarose can be used for situations requiring reversible binding and elution of avidin components.
Highlights:
- Enable purification or removal of avidin or streptavidin samples
- Resins are crosslinked beaded agarose onto which biotin or iminobiotin are covalently immobilized
- Use Biotin Agarose for non-reversible binding (e.g., removal of avidin components from a sample)
- Use Iminobiotin Agarose for reversible binding (e.g., non-denaturing purification of avidin)

Specifications
Pierce Iminobiotin Agarose | |
Pierce™ | |
Streptavidin | |
Biotin Derivative | |
Affinity, Agarose Resin |
Upon receipt store at 4°C. | |
5 mL | |
Agarose | |
Bottle |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.