missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Immobilized TCEP Disulfide Reducing Gel

Efficiently reduce protein or peptide disulfide bonds with this stable, odorless, beaded agarose resin on which Tris (2-carboxyethyl) phosphine is immobilized.

Brand:  Thermo Scientific™ 77712

Product Code. 10647344

  • £215.00 / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Immobilized TCEP Disulfide Reducing Gel is Tris(2-carboxyethyl)phosphine that is covalently immobilized to beaded agarose resin to enable efficient reduction of peptide and protein disulfide bonds.

Trialkylphosphines serve as excellent agents for the reduction of disulfide bonds in proteins, peptides and other disulfide bond-containing molecules and are relatively unreactive toward other functional groups. The trialkylphosphine TCEP is stable in aqueous solutions and does not undergo the rapid oxidation that often occurs with other reducing agents such as dithiotreitol (DTT) and β-mercaptoethanol (BME). TCEP does not interfere with common sulfhydryl-reactive reagents (e.g., maleimide crosslinkers). Nevertheless, many protocols require recovery of the reduced sample separate from the reducing agent. Immobilized TCEP Disulfide Reducing Gel eliminates the need to use laborious and inefficient gel filtration methods to separate the reduced sample from the reducing agent. Immobilized TCEP Disulfide Reducing Gel may be adapted conveniently to a variety of scales and formats (i.e. batch, spin column and gravity-flow column procedures).

Highlights:

  • Odorless: reduce proteins at your desktop; contributes to a healthier lab environment
  • Stable: virtually no loss of activity when stored refrigerated
  • Versatile: reduces peptides and proteins over a broad range of pH, salt, detergent and temperature conditions
  • Flexible: resin slurry provides flexibility for use in spin tube, column or batch modes
  • Time-saving: no soluble reductant removal saves time and conserves sample
  • Efficient: reverses both natural disulfide bonds and disulfide bonds artificially incorporated by crosslinking; excellent alternative for uncoupling crosslinked proteins or peptides by agents such as DSP and DTSSP
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Upon receipt store at 4°C. Do not freeze.
Liquid
Disulfide Reducing Reagent
Pierce™
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberOL194207 Certificate TypeCertificate of Analysis Date22/02/2025 Catalog Number77712
Lot NumberZL407165 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number77712
Lot NumberZJ401358 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number77712
Lot NumberZH391271 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number77712
Lot NumberZE381313 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number77712
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Immobilized TCEP Disulfide Reducing Gel > Immobilized TCEP Disulfide Reducing Gel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.