Learn More
Thermo Scientific™ LC-SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxy-(6-amidocaproate))
Crosslink amines to sulfhydryls with this long crosslinker composed of NHS-ester and maleimide groups at ends of a stabilizing cyclohexane-containing, 15-atom spacer arm.
Brand: Thermo Scientific™ 22362
217.55 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce LC-SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a long cyclohexane-stabilized spacer arm (16.2 Ås)
Highlights:
- Ideal for coupling enzymes to antibodies as both enzyme activity and antibody specificity can be preserved after coupling
- Cyclohexane-containing spacer stabilizes the maleimide group
- Useful for the preparation of stable maleimide-activated proteins
- Reactive groups: NHS ester and maleimide
- Reactive toward: amino and sulfhydryl groups

Specifications
Amine-Sulfhydryl | |
LC-SMCC | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Maleimide, NHS Ester | |
Solid | |
DMF, DMSO | |
No |
No | |
447.48 | |
16.2 Å | |
Yes | |
Pierce™ | |
Heterobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard | |
Crosslinker |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
56Hours
12Minutes
56Seconds
44New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.