Learn More
Thermo Scientific™ Pierce™ NeutrAvidin™ Agarose
Acrylamide and agarose beads and columns of specially-modified, pH-stabilized avidin; the low-cost alternative to streptavidin biotin-binding supports.
Brand: Thermo Scientific™ 29201
View more versions of this product
Description
Thermo Scientific™ Pierce NeutrAvidin Resins provide less nonspecific binding and cleaner results than avidin or streptavidin for many kinds of biotin affinity procedures.
Additional Information and Online Ordering
NeutrAvidin Agarose and UltraLink™ Resins are superior alternatives to avidin and streptavidin beads in many biotin-based affinity applications. NeutrAvidin Protein is a modified avidin derivative that combines several key features to provide a biotin-binding protein with exceptionally low nonspecific binding properties. Native avidin is heavily glycosylated, resulting in its high pI and nonspecific binding properties. By contrast NeutrAvidin Protein contains no carbohydrate and has a near-neutral isoelectric point (6.3), features that provide for very low nonspecific binding and broader utility in a variety of applications.NeutrAvidin Resins are prepared by covalently coupling NeutrAvidin Protein to the beaded agarose or UltraLink Biosupport using efficient and stable chemistries, resulting in NeutrAvidin Agarose and UltraLink Resins that are leach-resistant and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.
Highlights:
Low nonspecific binding – unlike avidin, NeutrAvidin Protein has no carbohydrates, eliminating nonspecific binding problems caused by sugars; unlike streptavidin, NeutrAvidin Protein has no Arg-Tyr-Asp sequence, which can be a source of binding to cell surface molecules
Versatile resins – agarose (polysaccharide-based) and UltraLink Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods
Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)
Superior performance – High Capacity Resin has higher binding capacity than best alternative supplier streptavidin products, allowing use of small amounts of resin for experiments
Item-Specific Features:
- Agarose Resins are crosslinked 6% beaded agarose
- UltraLink Resins are beaded acrylamide support (UltraLink Biosupport)
- Plus and High Capacity Resins have a higher loading of NeutrAvidin Protein than the regular forms
Recommended for:
Immunoprecipitation; Purifying proteins that bind to biotinylated ligands; Capturing biotinylated cell-surface proteins; Purifying biotinylated peptides

Specifications
Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature. | |
NeutrAvidin™, Pierce™ | |
Biotin-Tagged Molecule | |
NeutrAvidin Protein | |
Affinity, Agarose Resin |
45 to 165 μm | |
10 mL | |
Agarose | |
Bottle |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.