missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.

Brand:  Thermo Scientific™ A32959

277.40 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 15624189

  • £299.00 / Pack of 20

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Alert:

For Research Use Only. Not for use in diagnostic procedures.

This item is not returnable. View return policy

Description

Description

The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
* EDTA is present only in Product No. A32959.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protease and Phosphatase Inhibitor
Wet Ice
Pierce™
Protease Inhibitor, Phosphatase Inhibitor
1 Vial, Store at 2°C to 8°C
Solid
10 mL Solution
20 tablets
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZF4303992 Certificate TypeCertificate of Analysis Date20/06/2024 Catalog NumberA32959
Lot NumberZA4147963 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberA32959
Lot NumberYG3916413 Certificate TypeCertificate of Analysis Date05/08/2023 Catalog NumberA32959
Lot NumberYA3774002 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog NumberA32959
Lot NumberXF3579444 Certificate TypeCertificate of Analysis Date30/08/2022 Catalog NumberA32959
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets > 20 tablets; 1 tablet is suffcient for 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.