missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein Coated Plates

Thermo Scientific™ Pierce Protein A, G, A/G and L Coated Plates provide an alternative to direct, passive adsorption methods for immobilizing antibodies for ELISA and other plate-based assay techniques.

Brand:  Thermo Scientific™ 15155

 View more versions of this product

Product Code. 10298153

  • £246.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Standard Capacity, Affinity
Note: Plates are activated to 1 μL and supplied pre-blocked with Thermo Scientific SuperBlock Blocking Buffer. Use these plates for single-antibody assays only. In multiple antibody assays, such as sandwich ELISAs, the first antibody cannot block all binding sites and subsequent antibodies will bind to the plate, resulting in false positives.

Upon receipt store plates at 4°C in unopened pouches. Place opened, unused plates in a resealable bag and store desiccated at 4°C. Plates are shipped at ambient temperature.
Pierce
Detection Plate, Immunoassay, ELISA
96
5 Plates
5 plates
Standard
Fluorescence
Black
Polystyrene
Protein A
SuperBlock™ Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZE394138 Certificate TypeCertificate of Analysis Date04/06/2024 Catalog Number15155
Lot NumberVJ309199 Certificate TypeCertificate of Analysis Date22/10/2020 Catalog Number15155
Lot NumberTJ268699 Certificate TypeCertificate of Analysis Date16/11/2018 Catalog Number15155
Lot NumberTB263996 Certificate TypeCertificate of Analysis Date12/03/2018 Catalog Number15155
Lot NumberSG253515 Certificate TypeCertificate of Analysis Date31/08/2017 Catalog Number15155
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Protein Coated Plates > Protein A; Black, 96-well

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.