Learn More
Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl
Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.
£102.00 - £302.00
Specifications
Color | White |
---|---|
Product Type | Detergent |
Solubility | 10% (aq, w/v) solution must be clear, colorless to slightly yellow |
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C. |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11821475
|
Thermo Scientific™
28364 |
100 g |
£102.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10465715
|
Thermo Scientific™
28365 |
1 kg |
£302.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).
Additional Information and Online Ordering
This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).
Highlights:
- Popular anionic detergent for a variety of protein methods
- Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
- Common component of cell lysis buffers
Detergent properties (pure C12):
- Molecular Weight: 288.5
- Detergent Class: Ionic (anionic)
- Aggregation Number: 62
- Micelle Molecular Weight: 18,000
- Critical Micelle Concentration (CMC): 6 to 8mM (0.1728 to 0.2304%, w/v)
- Cloud Point: greater than 100°C
- Dialyzable: No
Storage Requirements
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
- Should be stored between the following temperatures: 20°C to 25°C
Specifications
White | |
10% (aq, w/v) solution must be clear, colorless to slightly yellow | |
Powder |
Detergent | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C. | |
Detergent (Pure) |
Safety and Handling
- sodium dodecyl sulphate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.