Learn More
Thermo Scientific™ Pierce™ Streptavidin Plus UltraLink™ Resin
Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.
£415.00 - £684.00
Specifications
Capacity (Metric) | 10 mg Biotinylated BSA Binding per mL Resin |
---|---|
Content And Storage | Upon receipt store at 4°C. Product is shipped at ambient temperature. |
Particle Size | 45 to 165 μm |
Product Line | Pierce™, UltraLink™ |
Protein Form | Recombinant |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
11889390
|
Thermo Scientific™
53116 |
2 mL | Binding ≥/= 48 μg Biotin/mL Resin (≥/= 4 mg Biotinylated BSA/mL Resin) |
£415.00
2mL |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
10259973
|
Thermo Scientific™
53117 |
5 mL | Binding ≥/= 48 μg Biotin/mL Resin (≥/= 4 mg Biotinylated BSA/mL Resin) |
£684.00
5mL |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
Description
Thermo Scientific™ Pierce Streptavidin Resins comprise a wide selection of high-quality affinity beads and columns for nearly any kind of biotin-based purification procedure.
Additional Information and Online Ordering »
Highlights:
Proven coupling chemistries – the methods used to prepare these resins are superior to typical cyanogen bromide (CNBr) immobilization, providing high functionality, low nonspecific binding and excellent leach-resistance
Versatile resins – Agarose (polysaccharide-based) and UltraLink™ Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods
Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)
Superior performance – High Capacity Resin has 2 to 3 times higher binding capacity than alternative supplier products, allowing use of small amounts of resin for experiments
Item-Specific Features:
- Agarose Resins are crosslinked 6% beaded agarose
- UltraLink Resins beaded acrylamide support (UltraLink Biosupport)
- Plus and High Capacity Resins have higher loading (i.e. density) of immobilized streptavidin protein than the regular forms
Recommended for:
Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies; Cell-surface labeling with biotinylation reagents, followed by precipitation with immobilized streptavidin; Purification of cell-surface glycoproteins using biotinylated Concanavalin A; Recovery of single-stranded DNA for dideoxy sequencing
Spécification
10 mg Biotinylated BSA Binding per mL Resin | |
45 to 165 μm | |
Recombinant | |
Resin | |
Binding ≥/= 48 μg Biotin/mL Resin (≥/= 4 mg Biotinylated BSA/mL Resin) | |
Affinity, UltraLink™ Resin |
Upon receipt store at 4°C. Product is shipped at ambient temperature. | |
Pierce™, UltraLink™ | |
Biotin-Tagged Molecule | |
Streptavidin | |
Bottle |
For Research Use Only. Not for use in diagnostic procedures.