Learn More
Thermo Scientific™ Pierce™ Tissue Strainers, 250 μm, 2.5 mL
Rapidly separate cells or cell lysate from tissue debris after mechanical disruption using a 250-micron mesh filter and gravity or gentle centrifugation.
Brand: Thermo Scientific™ 87791
160.55 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
The Thermo Scientific™ Pierce Tissue Strainer fits standard 15mL conical tubes for quick separation of cells or cell lysate from tissue debris through a 250μm mesh filter after mechanical disruption using gravity or gentle centrifugation.
The easy-to-use Pierce Tissue Strainers are made of strong nylon and enable rapid isolation cells or uniform protein extracts from up to 2.5mL of tissue sample per load. The tissue strainers fit into standard 15mL conical tubes, making the processing and recovery of samples simple and reliable.
Highlights:
Convenient remove tissue debris from tissue homogenate within minutes using gravity filtration or gentle centrifugation
Compatible unique design fits into standard 15mL conical tubes
Versitle remove debris from a variety of tissues types, including liver, heart, brain, kidney, lung and spleen
Recommended for:
Use with the Subcellular Protein Fractionation Kit for Tissues and other protein extraction reagent protocols that start with tissue samples; Improve uniformity of tissue homogenates to facilitate sample handling; Protect downstream instrumentation and devices by removing tissue clumps and debris that may foul injection ports and columns

Specifications
Tissue Strainer | |
250μM Nylon Mesh on Inert Plastic Frame | |
Pierce™ | |
50 Units |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.