missing translation for 'onlineSavingsMsg'
Learn More
Piperonyl butoxide, tech. 90%, Thermo Scientific Chemicals Product Code.: 15467268

Piperonyl butoxide, tech. 90%, Thermo Scientific Chemicals

Product Code. 15467268
25 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 15467268

Brand: Thermo Scientific Alfa Aesar H60771.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Piperonyl butoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonyl butoxide is used as a pharmaceutical intermediate.

Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-03-6
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
MDL Number MFCD00005842
InChI Key FIPWRIJSWJWJAI-UHFFFAOYSA-N
Synonym piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergistShow MoreShow Less
PubChem CID 5794
ChEBI CHEBI:32687
SMILES CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC

Specifications

Density 1.059
Boiling Point 155°C (0.3 mmHg)
Flash Point 171°C (340°F)
Refractive Index 1.498
Quantity 25 mL
UN Number UN3082
Beilstein 288063
Solubility Information Soluble in benzene,ethanol and methanol. Not miscible or difficult to mix with water.
IUPAC Name 5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
Formula Weight 338.44
Percent Purity 90%
Grade Technical
Chemical Name or Material Piperonyl butoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU11J016 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberH60771.AE,H60771.AC
Lot Number81401312 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60771.AE,H60771.AC
Lot Number81201627 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60771.AE,H60771.AC
Lot NumberW19E034 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60771.AE,H60771.AC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Piperonyl butoxide, tech. 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.