missing translation for 'onlineSavingsMsg'
Learn More
Pivaloyl hydrazide, 98+%, Thermo Scientific Chemicals Product Code.: 10525793

Pivaloyl hydrazide, 98+%, Thermo Scientific Chemicals

Product Code. 10525793
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10525793

Brand: Thermo Scientific Acros 377750050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42826-42-6
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.16
MDL Number MFCD01310839
InChI Key OARJXUPBZNUYBG-UHFFFAOYSA-N
Synonym 2,2-dimethylpropionic acid hydrazide, pivalic acid hydrazide, pivalohydrazide, 2,2-dimethylpropanohydrazide, pivaloylhydrazine, 2,2-dimethyl-propionic acid hydrazide, propanoic acid, 2,2-dimethyl-, hydrazide, pivalic acid, hydrazide, 2,2-dimethylpropionicacidhydrazide, trimethylacethydrazideShow MoreShow Less
PubChem CID 545147
IUPAC Name 2,2-dimethylpropanehydrazide
SMILES CC(C)(C)C(=O)NN

Specifications

Melting Point 65°C to 67°C
Color White
Boiling Point 95°C to 110°C (1.0 mbar)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC(O)NHNH2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohols
Formula Weight 116.16
Percent Purity 98+%
Physical Form Crystals
Chemical Name or Material Pivaloyl hydrazide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0374067 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number377750010,377750050
compliance-icons compliance-icons
Product Identifier
  • Pivaloyl hydrazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pivaloyl hydrazide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.