Learn More
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock
HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.
Brand: Fisherbrand™ 05 36 2400
36.29 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- Plastic Disposable Syringe
- Polypropylene
- Luer Lock
- Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
- No silicone or oil lubricant is required in the barrel

Specifications
Polypropylene | |
5 mL | |
Luer Lock |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.