missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Replacement Electrode Wire
Replacement Electrode Wire is an accessoty for use with Verti-Gel Maxi, 2-Gel and 4-Gel Electrophoresis System.
Brand: Fisherbrand™ RPW0.2
Description
- Made of platinum
Specifications
Major types of 80 x 100 mm and 100 x 100 mm Precast Gel | |
Electrode Wire Replacement | |
Platinum | |
500 x 0.2 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Replacement Electrode Wire > 500 x 0.2mm
Spot an opportunity for improvement?Share a Content Correction