Learn More
Molecular Probes™ Pluronic™ F-127 (20% Solution in DMSO)
Pluronic™ F-127 (20% Solution in DMSO)
Brand: Molecular Probes™ P3000MP
52.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
Pluronic F-127 *20% solution in DMSO* is a nonionic, surfactant polyol (molecular weight approximately 12,500 daltons) that has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media. Pluronic F-127 is commonly used to help disperse the acetoxymethyl (AM) esters of our ion indicators as well as our cell tracer dyes such as CFDA-SE.
Specifications
Liquid | |
1 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2935433 | Certificate of Analysis | 19/08/2024 | P3000MP |
2714438 | Certificate of Analysis | 16/08/2023 | P3000MP |
2510678 | Certificate of Analysis | 03/08/2022 | P3000MP |
2339925 | Certificate of Analysis | 16/08/2021 | P3000MP |
2291566 | Certificate of Analysis | 30/12/2020 | P3000MP |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.