missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Poly(A) Tail-Length Assay Kit

This kit utilizes a novel G/I extension technique which preserves the 3' end of RNAs and ensures accurate measurement of poly(A) tail-length. And, all reaction components have been optimized so you get quality data quickly.

Brand:  Invitrogen™ 764551KT

Product Code. 15774678

  • £562.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Sufficient reagents for 5 G/I tailing, 20 reverse transcription, and 80 PCR reactions.

HeLa Total RNA and Control Human Actin PCR Forward Primer to verify components and protocol.

Store reagents at -15°C to -30°C in a non-frost-free freezer.
Store HeLa Total RNA at -80°C.
Poly(A) RNA Polymerase
Total Eukaryotic RNA (100 ng to 2 μg)
1.25 U/μL
Dry Ice
Poly(A) tail-length detection
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2791363 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number764551KT
Lot Number2812421 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number764551KT
Lot Number2814321 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number764551KT
Lot Number2821279 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number764551KT
Lot Number2795511 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number764551KT
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Poly(A) Tail-Length Assay Kit > 1 Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.