missing translation for 'onlineSavingsMsg'
Learn More
Logistik™ Women's FItted Polycotton Laboratory Coat Product Code.: 13102180

Logistik™ Women's FItted Polycotton Laboratory Coat

Product Code. 13102180
100 cm, 39.3 in.
Click to view available options
Size (Metric) Chest:
92 cm
96 cm
100 cm
104 cm
108 cm
128 cm
Size (English) Chest:
36.2 in.
37.8 in.
39.3 in.
40.9 in.
42.5 in.
50.4 in.
This item is not returnable. View return policy

Product Code. 13102180

Brand: Logistik™ 607V3RB100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Logistik Unicorp™ Women's FItted Polycotton Laboratory Coat is a classically designed, comfortable, fitted laboratory coat suitable for a range of applications.

  • Gently contoured fit for optimum comfort
  • Includes two lower patch pockets, breast pocket, concealed stud fastened front, stud fastened cuffs and a generous 107cm length
  • Not CE marked

Specifications

Product Type General Purpose Lab Coat
Size Small/Medium
Gender Women's
Material Polycotton
Color Royal Blue
Cuff Style Stud-fastened
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Description Size 16
Size (English) Chest 39.3 in.
Size (Metric) Chest 100 cm
Includes Lower patch pockets, breast pocket, concealed stud fastened front, stud fastened cuffs
Length (Metric) 107 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Logistik™ Women's FItted Polycotton Laboratory Coat >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.