Learn More
Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials
Fit securely into place to seal glass specimen vials. Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials are available in a variety of sizes specifically designed to work with Fisherbrand™ Push-in Cap Glass Specimen Vials.
Brand:  Fisherbrand™ 15354789
 View more versions of this product
Product Code. 15354789
  • £44.10 / Pack of 300
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Specifically designed for use with Fisherbrand™ Push-in Cap Glass Specimen Vials
  • Polyethylene construction, natural color
  • Available in a variety of sizes to match vial specifications
  • Push-in cap specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Natural
Polyethylene
Push-in Cap
22 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials > Cap Size: 22mm