missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Conical Ground Joint Stopper
£12.75 - £30.25
Specifications
Description | Conical bottom |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Type | Ground Joint Stopper |
Shape | Square |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15567831
|
BRAND™
144425 |
NS 19/26 |
£12.85
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15547831
|
BRAND™
144415 |
NS 12/21 |
£19.15
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15557831
|
BRAND™
144420 |
NS 14/23 |
£16.85
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15577831
|
BRAND™
144430 |
NS 24/29 |
£18.80
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15537831
|
BRAND™
144410 |
NS 10/19 |
£17.35
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15587831
|
BRAND™
144435 |
NS 29/32 |
£30.25
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15527831
|
BRAND™
144405 |
NS 7/16 |
£12.75
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
Specifications
Conical bottom | |
Polypropylene | |
Square |
Autoclavable | |
Ground Joint Stopper | |
1/Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Polypropylene Conical Ground Joint Stopper