Learn More
BRAND™ Polypropylene Conical Ground Joint Stopper
Brand:  BRAND™ 144430
13.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15577831
  • £18.80 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Conical bottom
NS 24/29
Polypropylene
Square
1/Ea.
Autoclavable
Ground Joint Stopper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Polypropylene Conical Ground Joint Stopper > Size: 24/29 NS