Learn More
BRAND™ Polypropylene Conical Ground Joint Stopper
Brand: BRAND™ 144410
9.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Conical bottom | |
NS 10/19 | |
Polypropylene | |
Square |
1/Ea. | |
Autoclavable | |
Ground Joint Stopper |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ Polypropylene Conical Ground Joint Stopper > Size: 10/19 NS
Spot an opportunity for improvement?Share a Content Correction