Learn More
Brand™ Polypropylene Funnel Holders
To support 2 funnels with top outer diameter of 50 to 120mm.
Brand: Brand™ 148502
Description
- Easily mounted on support rods with an outer diameter of 8 to 14mm
- Includes locking nut
- Provided in packs of 5

Specifications
Locking Nut | |
Polypropylene | |
2 funnels | |
5/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.