missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microcentrifuge Tubes

Use these tubes for high speed centrifugation applications. Fisherbrand™ Polypropylene Microcentrifuge Tubes feature a frosted writing surface on closure and side of tube (except for natural tubes) which allows for easy and convenient sample identification.

Brand:  Fisherbrand™ FB74111

38.17 GBP valid until 2025-12-31
Use promo code "24328" to get your promotional price.


 View more versions of this product

Product Code. 11558252

  • £58.48 / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Graduated polypropylene tubes
  • Suitable for withstanding the stress of high speed centrifugation up to 20,000xg
  • One-piece construction incorporates a snug fitting and reliable cap and the bottom is reinforced for added protection against leakage
  • Autoclavable
Specifications

Specifications

Polypropylene
2 mL
Polypropylene
1000/Pk.
Yes
Autoclavable
Natural
Microcentrifuge
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Polypropylene Microcentrifuge Tubes > Capacity: 2mL; Color: Natural

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.