missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene, Spare Screw-on Tap Product Code.: 12390449
In Stock
to see stock.
Add to basket

Polyethylene, Spare Screw-on Tap

Product Code. 12390449
Aspirator ASP-520 Series
Click to view available options
For Use With (Equipment):
Aspirator ASP-520 Series

Product Code. 12390449

Brand: S Murray SAMCO P023/04

In Stock
to see stock.
Edge
Add to basket
Add to basket

For Use With: Aspirator ASP-520 Series

  • High density polythene tap
  • For use with 5 and 10mL aspirators (PN: P023/01 and P023/02)
  • Tap assembly increases aspirator width by 35mm

Specifications

For Use With (Equipment) Aspirator ASP-520 Series
Material Polyethylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene, Spare Screw-on Tap >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.