missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Medium Form Melting Crucibles
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ L 79MF/8
This item is not returnable.
View return policy
Description
- Available in a variety of sizes
- Sizes up to 175mL glazed except rim
- 1,000mL and larger are glazed except outside base
- Complies with DIN VDE 0335, type C 100
Specifications
Porcelain | |
10 mL | |
25mm |
Glazed Inside and Outside Except Base | |
30mm | |
Melting Crucible |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haldenwanger™ Porcelain Medium Form Melting Crucibles > Capacity: 10mL; Height: 25mm; Glazed Inside and Outside Except Base
Spot an opportunity for improvement?Share a Content Correction