Learn More
Thermo Scientific™ POROS™ HQ50 RoboColumn
RoboColumn™ chromatography products are small columns provided in 8-column strips.
Brand: Thermo Scientific™ A30717
141.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : CAS Number : 64-17-5 Weight : 0.05000kg
Description
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS™ HQ50 resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS HQ is a high-performance chromatography resin based on a quaternized polyethyleneimine functional group, yielding a high capacity. It is designed for the separation and purification of biomolecules for analytical to process scale separations
Features of the resin:
- Higher productivity—through high throughput and high dynamic capacity associated with high performance chromatography
- Chemical stability—allows aggressive cleaning and sanitization
- Enhanced biomolecule access—provided via large pores (500-10,000 Å)
- Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
- Better separation methods in less time—the speed of high performance chromatography reduces weeks of experimentation to only a few hours, leaving more time to explore all the variables of your separation
RoboColumn is a registered trademark of ATOLL GmbH.
Specifications
Anion Exchange Column | |
200 μL | |
Anion Exchange, Protein Purification |
Store at 2°C to 8°C or at room temperature. | |
16 to 1000 cm/hr. | |
116 psi (8 bar) |