Learn More
Fisherbrand™ Portable Balance
Portable Balance with a slim, stackable design provides basic weighing functionality and competitive performance.
£48.05 - £75.90
Specifications
Display Type LCD
For Use With (Application) Weighing
Includes 3 AA (LR6) Batteries
Stabilization Time 2.000 sec.
Temperature Range (English) 64°F to 77°F
View More Specs
Products 4
Product Code Brand Capacity Model Price Quantity & Availability  
Product Code Brand Capacity Model Price Quantity & Availability  
15939345
View Documents Promotion Details
Fisherbrand™
30467785
220 g FCS221
£48.05
Each
15949345
View Documents Promotion Details
Fisherbrand™
30467786
620 g FCS621
£59.82
Each
15909355
View Documents Promotion Details
Fisherbrand™
30467973
2200 g FCS2200
£48.50
Each
15919355
View Documents Promotion Details
Fisherbrand™
30467974
5200 g FCS5200
£75.90
Each
Description
Description
  • Intuitive two-button design makes for easy operation
  • Slim, space-saving design allows for stackable storage
  • Enlarged weighing platform can adequately handle large samples and a variety of objects
Specifications
Specifications
LCD
3 AA (LR6) Batteries
64°F to 77°F
2 Years
5 in.
5.2 in.
0.9 lb.
Weighing
2.000 sec.
18°C to 25°C
205 x 140 x 41 mm (8.1 x 5.5 x 1.6 in.)
125 mm
132 mm
0.4 kg
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Portable Balance