Learn More
Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals
£16.58 - £176.88
Chemical Identifiers
CAS | 16919-27-0 |
---|---|
Molecular Formula | F6K2Ti |
MDL Number | MFCD00011380 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414303
|
Thermo Scientific Alfa Aesar
A10915.22 |
100 g |
£16.58
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424303
|
Thermo Scientific Alfa Aesar
A10915.36 |
500 g |
£49.45
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434303
|
Thermo Scientific Alfa Aesar
A10915.0E |
2500 g |
£176.88
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.
Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
16919-27-0 | |
F6K2Ti | |
MFCD00011380 |
Specifications
235°C to 237°C | |
F6K2Ti | |
MFCD00011380 | |
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia. | |
97% |
16919-27-0 | |
K2TiF6 | |
UN3288 | |
240.07 | |
Potassium hexafluorotitanate |
Safety and Handling
GHS H Statement
H318-H302-H317-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 240-969-9
missing translation for 'rtecsNumber' : TT1575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only